missing translation for 'onlineSavingsMsg'
Learn More

1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-piperidinecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7129975
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
F09141G 1 g
F09145G 5 g
2 options

Catalog No. F09141G

Supplier: TCI America F09141G

Only null left

Chemical Identifiers

CAS 148928-15-8
Molecular Formula C21H21NO4
Molecular Weight (g/mol) 351.402
MDL Number MFCD00273475
InChI Key OJYOOHSQOIDDOO-UHFFFAOYSA-N
Synonym fmoc-inp-oh, fmoc-isonipecotic acid, 1-fmoc-piperidine-4-carboxylic acid, 1-9h-fluoren-9-ylmethoxy carbonyl piperidine-4-carboxylic acid, n-fmoc-isonipecotic acid, fmoc-piperidine-4-carboxylic acid, n-fmoc-4-piperidinecarboxylic acid, 1,4-piperidinedicarboxylicacid, 1-9h-fluoren-9-ylmethyl ester, n-9-fluorenylmethyloxycarbonyl-isonipecotic acid, 1-9h-fluoren-9-ylmethoxycarbonyl piperidine-4-carboxylic acid
PubChem CID 2736490
IUPAC Name 1-(9H-fluoren-9-ylmethoxycarbonyl)piperidine-4-carboxylic acid
SMILES C1CN(CCC1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 1 g
Formula Weight 351.40
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-piperidinecarboxylic Acid