missing translation for 'onlineSavingsMsg'
Learn More

1,9-Nonanediol, 97%

Catalog No. AAL0633622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0633622 100 g
AAL0633614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0633622 Supplier Thermo Scientific Chemicals Supplier No. L0633622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3937-56-2 | C9H20O2 | 160.257 g/mol

1,9-Nonanediol is used in organic synthesis. It also can react with 3,4-dihydro-2H-pyran and 9-(tetrahydropyrane-2-yloxy)nonan-1-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,9-Nonanediol is used in organic synthesis. It also can react with 3,4-dihydro-2H-pyran and 9-(tetrahydropyrane-2-yloxy)nonan-1-ol.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.

Chemical Identifiers

CAS 3937-56-2
Molecular Formula C9H20O2
Molecular Weight (g/mol) 160.257
MDL Number MFCD00002991
InChI Key ALVZNPYWJMLXKV-UHFFFAOYSA-N
Synonym 1,9-nonanediol, 1,9-dihydroxynonane, .alpha.,.omega.-nonanediol, nonamethylene glycol, alpha,omega-nonanediol, 1,9-nonandiol, 1,9-nonane-diol, nonan-1,9-diol, acmc-209j4x
PubChem CID 19835
IUPAC Name nonane-1,9-diol
SMILES C(CCCCO)CCCCO

Specifications

Melting Point 45°C to 48°C
Density 0.95
Boiling Point 288°C
Flash Point 162°C (323°F)
Quantity 100 g
Beilstein 1737531
Solubility Information Soluble in methanol.
Formula Weight 160.26
Percent Purity 97%
Chemical Name or Material 1,9-Nonanediol
EINECSNumber 223-517-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only