missing translation for 'onlineSavingsMsg'
Learn More

1,9-Dibromononane, 97%, Thermo Scientific™

$179.48 - $179.48

Chemical Identifiers

CAS 4549-33-1
Molecular Formula C9H18Br2
Molecular Weight (g/mol) 286.04
InChI Key WGAXVZXBFBHLMC-UHFFFAOYSA-N
Synonym nonane, 1,9-dibromo, nonamethylene dibromide, nonamethylene bromide, 1,9-dibromnonane, nonane,9-dibromo, 1,9-dibromononane, pubchem3900, acmc-1ai7y, ksc236g2b
PubChem CID 20677
IUPAC Name 1,9-dibromononane
SMILES C(CCCCBr)CCCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC112870050
SDS View Documents
Thermo Scientific Chemicals
112870050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC112870250
SDS View Documents
Thermo Scientific Chemicals
112870250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4549-33-1
286.04
nonane, 1,9-dibromo, nonamethylene dibromide, nonamethylene bromide, 1,9-dibromnonane, nonane,9-dibromo, 1,9-dibromononane, pubchem3900, acmc-1ai7y, ksc236g2b
1,9-dibromononane
C9H18Br2
WGAXVZXBFBHLMC-UHFFFAOYSA-N
20677
C(CCCCBr)CCCCBr

Specifications

4549-33-1
100.0
285°C to 288°C
Authentic
Glass bottle
1.4950 to 1.497
5g
1.4
WGAXVZXBFBHLMC-UHFFFAOYSA-N
1,9-dibromononane
20677
97%
96.0
1.4000g/mL
>112°C
96% min. (GC)
C9H18Br2
Br(CH2)9Br
01, 166
nonane, 1,9-dibromo, nonamethylene dibromide, nonamethylene bromide, 1,9-dibromnonane, nonane,9-dibromo, 1,9-dibromononane, pubchem3900, acmc-1ai7y, ksc236g2b
C(CCCCBr)CCCCBr
286.04
286.04
1, 9-Dibromononane, 97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 224-913-0

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only