missing translation for 'onlineSavingsMsg'
Learn More

1,9-Dibromononane, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 112870050

 View more versions of this product

Catalog No. AC112870050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1, 9-Dibromononane
4549-33-1
100.0
285°C to 288°C
Authentic
Glass bottle
1.4950 to 1.497
5g
1.4
WGAXVZXBFBHLMC-UHFFFAOYSA-N
1,9-dibromononane
20677
97%
97%
96.0
1.4000g/mL
>112°C
96% min. (GC)
C9H18Br2
Br(CH2)9Br
01, 166
nonane, 1,9-dibromo, nonamethylene dibromide, nonamethylene bromide, 1,9-dibromnonane, nonane,9-dibromo, 1,9-dibromononane, pubchem3900, acmc-1ai7y, ksc236g2b
C(CCCCBr)CCCCBr
286.04
286.04

Chemical Identifiers

4549-33-1
286.04
nonane, 1,9-dibromo, nonamethylene dibromide, nonamethylene bromide, 1,9-dibromnonane, nonane,9-dibromo, 1,9-dibromononane, pubchem3900, acmc-1ai7y, ksc236g2b
1,9-dibromononane
C9H18Br2
WGAXVZXBFBHLMC-UHFFFAOYSA-N
20677
C(CCCCBr)CCCCBr
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 224-913-0

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only