missing translation for 'onlineSavingsMsg'
Learn More

1,9-Dibromononane, 97%

Catalog No. AAA1432409
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1432409 10 g
AAA1432414 25 g
AAA1432418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1432409 Supplier Thermo Scientific Chemicals Supplier No. A1432409

CAS: 4549-33-1 | C9H18Br2 | 286.051 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4549-33-1
Molecular Formula C9H18Br2
Molecular Weight (g/mol) 286.051
MDL Number MFCD00000279
InChI Key WGAXVZXBFBHLMC-UHFFFAOYSA-N
Synonym nonane, 1,9-dibromo, nonamethylene dibromide, nonamethylene bromide, 1,9-dibromnonane, nonane,9-dibromo, 1,9-dibromononane, pubchem3900, acmc-1ai7y, ksc236g2b
PubChem CID 20677
IUPAC Name 1,9-dibromononane
SMILES C(CCCCBr)CCCCBr

Specifications

Density 1.41
Boiling Point 285°C to 288°C
Flash Point >110°C (230°F)
Refractive Index 1.4965
Linear Formula Br(CH2)9Br
Quantity 10 g
Beilstein 1737598
Formula Weight 286.06
Percent Purity 97%
Chemical Name or Material 1,9-Dibromononane
EINECSNumber 224-913-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only