missing translation for 'onlineSavingsMsg'
Learn More

1,8-Octanedithiol, 98%

Catalog No. AAB2469303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2469303 1 g
AAB2469306 5 g
AAB2469314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2469303 Supplier Thermo Scientific Chemicals Supplier No. B2469303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1191-62-4 | C8H18S2 | 178.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1191-62-4
Molecular Formula C8H18S2
Molecular Weight (g/mol) 178.35
MDL Number MFCD00003574
InChI Key PGTWZHXOSWQKCY-UHFFFAOYSA-N
Synonym 1,8-octanedithiol, 1,8-dimercaptooctane, octamethylene dimercaptan, 1,8-octamethylenedithiol, unii-kh3w3161hs, fema no. 3514, 1,8-octane dithiol, nanothinks tm thio8, acmc-209a1j, dsstox_cid_27474
PubChem CID 14493
IUPAC Name octane-1,8-dithiol
SMILES SCCCCCCCCS

Specifications

Density 0.96
Boiling Point 269°C to 270°C
Flash Point 113°C (235°F)
Odor Unpleasant
Refractive Index 1.505
Quantity 1 g
Beilstein 1735431
Sensitivity Air Sensitive
Formula Weight 178.36
Percent Purity 98%
Chemical Name or Material 1,8-Octanedithiol
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 214-738-8
RTECSNumber RG9610000
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only