missing translation for 'onlineSavingsMsg'
Learn More

1,8-Naphthalic anhydride, 97%

CAS: 81-84-5 | C12H6O3 | 198.18 g/mol

$180.30 - $180.30

Chemical Identifiers

CAS 81-84-5
Molecular Formula C12H6O3
Molecular Weight (g/mol) 198.18
MDL Number MFCD00006925
InChI Key GRSMWKLPSNHDHA-UHFFFAOYSA-N
Synonym 1,8-naphthalic anhydride, benzo de isochromene-1,3-dione, naphthalic anhydride, protect, naphthalic acid anhydride, 1,8-naphthalic acid anhydride, pakarli, protect agrochemical, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 1h,3h-benzo de isochromene-1,3-dione
PubChem CID 6693
ChEBI CHEBI:82246
IUPAC Name 3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
SMILES O=C1OC(=O)C2=C3C(C=CC=C13)=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC128145000
SDS View Documents
Thermo Scientific Chemicals
128145000
500 g
Each for $180.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

81-84-5
198.18
GRSMWKLPSNHDHA-UHFFFAOYSA-N
6693
3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
C12H6O3
MFCD00006925
1,8-naphthalic anhydride, benzo de isochromene-1,3-dione, naphthalic anhydride, protect, naphthalic acid anhydride, 1,8-naphthalic acid anhydride, pakarli, protect agrochemical, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 1h,3h-benzo de isochromene-1,3-dione
CHEBI:82246
O=C1OC(=O)C2=C3C(C=CC=C13)=CC=C2

Specifications

81-84-5
100.0
Beige to Brown
0.5% max.
97%
C12H6O3
500 g
01,713
Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,soluble in acetic acid
O=C1OC(=O)C2=C3C(C=CC=C13)=CC=C2
198.18
CHEBI:82246
97%
1, 8-Naphthalic anhydride, 97%
96.0
>269.0°C
272°C
Authentic
Plastic bottle
MFCD00006925
17, 521
1,8-naphthalic anhydride, benzo de isochromene-1,3-dione, naphthalic anhydride, protect, naphthalic acid anhydride, 1,8-naphthalic acid anhydride, pakarli, protect agrochemical, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 1h,3h-benzo de isochromene-1,3-dione
GRSMWKLPSNHDHA-UHFFFAOYSA-N
3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
6693
198.18
Powder
Videos
Safety and Handling

Safety and Handling

MOISTURE SENSITIVE

EINECSNumber : 201-380-2

RTECSNumber : QK5350000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only