missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dinitronaphthalene 85.0+%, TCI America™

Catalog No. D0837500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0837500G 500 g
D083725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0837500G Supplier TCI America Supplier No. D0837500G
Only null left

Chemical Identifiers

CAS 602-38-0
Molecular Formula C10H6N2O4
Molecular Weight (g/mol) 218.168
MDL Number MFCD00003918
InChI Key AVCSMMMOCOTIHF-UHFFFAOYSA-N
PubChem CID 11764
ChEBI CHEBI:50642
IUPAC Name 1,8-dinitronaphthalene
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CC=C2)[N+](=O)[O-]

Specifications

Melting Point 161°C
Color Yellow
Quantity 500 g
Synonym naphthalene, 1,8-dinitro, unii-h6d9q0k9e6, ccris 2840, 1,8-dinitro-naphthalen, naphthalene,8-dinitro, 1,8-dinitro naphthalene, 1,8-dinitro-naphthalene, dsstox_cid_5166, naphthalene,1,8-dinitro, acmc-1b07g
Formula Weight 218.17
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Dinitronaphthalene
EINECSNumber (4)-0325
RTECSNumber QJ4552000
TSCA Yes