missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diiodooctane (stabilized with Copper chip) 95.0+%, TCI America™

Catalog No. p-7127142
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21585G 5 g
D215825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21585G Supplier TCI America Supplier No. D21585G
Only null left

Chemical Identifiers

CAS 24772-63-2
Molecular Formula C8H16I2
Molecular Weight (g/mol) 366.025
MDL Number MFCD00001106
InChI Key KZDTZHQLABJVLE-UHFFFAOYSA-N
PubChem CID 90605
IUPAC Name 1,8-diiodooctane
SMILES C(CCCCI)CCCI

Specifications

Boiling Point 144°C
Quantity 5 g
Synonym octane, 1,8-diiodo, octamethylene diiodide, pubchem3851, octane,1,8-diiodo, acmc-1cljq, 1,8-bis iodanyl octane, 1,8-diiodooctane, stabilized with copper, stabilizedwithcopperchip, 1,8-diiodooctane, contains copper as stabilizer
Formula Weight 366.02
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,8-Diiodooctane (stabilized with Copper chip)