accessibility menu, dialog, popup

UserName

CAS RN 24772-63-2

IUPAC Name: 1,8-diiodooctane
Synonyms: 1,8-diiodooctane
Molecular Weight (g/mol): 366.03
Molecular Formula: C8H16I2
InChi Key: KZDTZHQLABJVLE-UHFFFAOYSA-N
SMILES: ICCCCCCCCI
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)