missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diiodooctane, 97+%, stab. with copper

Catalog No. AAA1086718
Change view
Click to view available options
Quantity:
25 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1086718 50 g
AAA1086714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1086718 Supplier Thermo Scientific Chemicals Supplier No. A1086718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24772-63-2 | C8H16I2 | 366.025 g/mol

1,8-Diiodooctane is utilized to improve the power conversion efficiency of polymer solar cells. It is also employed as a processing additive to improve the morphology and the efficiency of bulk heterojunctions solar cells based on the regioregular poly(3-hexylthiophene).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diiodooctane is utilized to improve the power conversion efficiency of polymer solar cells. It is also employed as a processing additive to improve the morphology and the efficiency of bulk heterojunctions solar cells based on the regioregular poly(3-hexylthiophene).

Solubility
Miscible with methanol.

Notes
Light sensitive. Incompatible with water and strong bases.

Chemical Identifiers

CAS 24772-63-2
Molecular Formula C8H16I2
MDL Number MFCD00001106
InChI Key KZDTZHQLABJVLE-UHFFFAOYSA-N
PubChem CID 90605
IUPAC Name 1,8-diiodooctane

Specifications

Density 1.84
Boiling Point 167°C to 169°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5635
Quantity 50 g
Beilstein 1735437
Sensitivity Light sensitive
Synonym octane, 1,8-diiodo, octamethylene diiodide, pubchem3851, octane,1,8-diiodo, acmc-1cljq, 1,8-bis iodanyl octane, 1,8-diiodooctane, stabilized with copper, stabilizedwithcopperchip, 1,8-diiodooctane, contains copper as stabilizer
Solubility Information Miscible with methanol.
SMILES C(CCCCI)CCCI
Molecular Weight (g/mol) 366.025
Formula Weight 366.03
Percent Purity ≥97%
Chemical Name or Material 1,8-Diiodooctane, Stabilized with copper
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 246-456-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only