missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diiodoanthracene 98.0+%, TCI America™

Catalog No. D41511G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41511G 1 g
D41515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41511G Supplier TCI America Supplier No. D41511G
Only null left

Chemical Identifiers

CAS 189105-78-0
Molecular Formula C14H8I2
Molecular Weight (g/mol) 430.027
InChI Key BBKOQZVGDMYPGV-UHFFFAOYSA-N
PubChem CID 10836367
IUPAC Name 1,8-diiodoanthracene
SMILES C1=CC2=CC3=C(C=C2C(=C1)I)C(=CC=C3)I

Specifications

Melting Point 201°C
Quantity 1 g
Formula Weight 430.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Diiodoanthracene
TSCA No
Recommended Storage Freezer