missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Dichlorohexadecafluorooctane 98.0+%, TCI America™
Supplier: TCI America D280425G
Specifications
| 1,8-Dichlorohexadecafluorooctane | |
| Colorless | |
| MFCD00155747 | |
| 1,8-Dichloroperfluorooctane | |
| FC(F)(Cl)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Cl | |
| 470.96 | |
| CHEBI:38872 | |
| ≥98.0% (GC) |
| 647-25-6 | |
| C8Cl2F16 | |
| 25 g | |
| GFQXWORJCNTDPU-UHFFFAOYSA-N | |
| 1,8-dichloro-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-hexadecafluorooctane | |
| 2774033 | |
| 470.96 | |
| Liquid |
Chemical Identifiers
| 647-25-6 | |
| 470.96 | |
| GFQXWORJCNTDPU-UHFFFAOYSA-N | |
| 2774033 | |
| 1,8-dichloro-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-hexadecafluorooctane |
| C8Cl2F16 | |
| MFCD00155747 | |
| 1,8-Dichloroperfluorooctane | |
| CHEBI:38872 | |
| FC(F)(Cl)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Cl |
Safety and Handling
TSCA : No