accessibility menu, dialog, popup

UserName

CAS RN 647-25-6

IUPAC Name: 1,8-dichloro-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-hexadecafluorooctane
Synonyms: pfdco 1,8-dichloro-perfluorooctane
Molecular Weight (g/mol): 470.96
Molecular Formula: C8Cl2F16
InChi Key: GFQXWORJCNTDPU-UHFFFAOYSA-N
SMILES: FC(F)(Cl)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results