missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dibromooctane 97.0+%, TCI America™

Catalog No. D1064500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1064500G 500 g
D106425G 25 g
D1064100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1064500G Supplier TCI America Supplier No. D1064500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4549-32-0
Molecular Formula C8H16Br2
Molecular Weight (g/mol) 272.024
MDL Number MFCD00000277
InChI Key DKEGCUDAFWNSSO-UHFFFAOYSA-N
Synonym octamethylene dibromide, octane, 1,8-dibromo, octamethylene bromide, 1,8-dibromo-octane, labotest-bb ltbb002638, 1,8-dibromoctane, octane,8-dibromo, dibromo-1,8 octane, pubchem3905, acmc-209k2r
PubChem CID 78310
IUPAC Name 1,8-dibromooctane
SMILES C(CCCCBr)CCCBr

Specifications

Color Yellow
Boiling Point 272°C
Quantity 500 g
Formula Weight 272.02
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,8-Dibromooctane