missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dibromooctane, 98%

Catalog No. AAA1565122
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1565122 100 g
AAA1565114 25 g
AAA1565118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1565122 Supplier Thermo Scientific Chemicals Supplier No. A1565122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4549-32-0 | C8H16Br2 | 272.024 g/mol

1,8-Dibromooctane is used as an additive in the preparation poly[[4,8-bis[(2-ethylhexyl)oxy]benzo[1,2-b:4,5-b']dithiophene-2,6-diyl][3-fluoro-2-[(2-ethylhexyl)carbonyl]thieno[3,4-b]thiophenediyl]]:[6,6]-phenylC71-butyric acid methyl ester, which finds application in the fabrication of organic photovoltaics (OPVs). Further, it is used in the preparation of (1,4-bis-(8-bromooctyloxy)-benzene).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Dibromooctane is used as an additive in the preparation poly[[4,8-bis[(2-ethylhexyl)oxy]benzo[1,2-b:4,5-b′]dithiophene-2,6-diyl][3-fluoro-2-[(2-ethylhexyl)carbonyl]thieno[3,4-b]thiophenediyl]]:[6,6]-phenylC71-butyric acid methyl ester, which finds application in the fabrication of organic photovoltaics (OPVs). Further, it is used in the preparation of (1,4-bis-(8-bromooctyloxy)-benzene).

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 4549-32-0
Molecular Formula C8H16Br2
MDL Number MFCD00000277
InChI Key DKEGCUDAFWNSSO-UHFFFAOYSA-N
Synonym octamethylene dibromide, octane, 1,8-dibromo, octamethylene bromide, 1,8-dibromo-octane, labotest-bb ltbb002638, 1,8-dibromoctane, octane,8-dibromo, dibromo-1,8 octane, pubchem3905, acmc-209k2r
PubChem CID 78310
IUPAC Name 1,8-dibromooctane

Specifications

Melting Point 15°C to 16°C
Density 1.468
Boiling Point 270°C to 272°C
Flash Point >110°C (230°F)
Refractive Index 1.4995
Quantity 100 g
Beilstein 1698114
Solubility Information Immiscible with water.
SMILES C(CCCCBr)CCCBr
Molecular Weight (g/mol) 272.024
Formula Weight 272.02
Percent Purity 98%
Chemical Name or Material 1,8-Dibromooctane
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 224-912-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only