missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Dibromoanthracene 98.0+%, TCI America™
$290.79 - $1,004.73
Chemical Identifiers
| CAS | 131276-24-9 |
|---|---|
| Molecular Formula | C14H8Br2 |
| Molecular Weight (g/mol) | 336.026 |
| InChI Key | JQQFVBWHOMXEEA-UHFFFAOYSA-N |
| PubChem CID | 630683 |
| IUPAC Name | 1,8-dibromoanthracene |
| SMILES | C1=CC2=CC3=C(C=C2C(=C1)Br)C(=CC=C3)Br |
Chemical Identifiers
| 131276-24-9 | |
| 336.026 | |
| 630683 | |
| C1=CC2=CC3=C(C=C2C(=C1)Br)C(=CC=C3)Br |
| C14H8Br2 | |
| JQQFVBWHOMXEEA-UHFFFAOYSA-N | |
| 1,8-dibromoanthracene |
Specifications
| 131276-24-9 | |
| Yellow | |
| 1 g | |
| C1=CC2=CC3=C(C=C2C(=C1)Br)C(=CC=C3)Br | |
| 336.026 | |
| 336.03 | |
| Crystalline Powder |
| 168°C | |
| C14H8Br2 | |
| JQQFVBWHOMXEEA-UHFFFAOYSA-N | |
| 1,8-dibromoanthracene | |
| 630683 | |
| ≥98.0% (GC) | |
| 1,8-Dibromoanthracene |
Safety and Handling
TSCA : No