missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diazabicyclo[5.4.0]-7-undecene Hydrogen Tribromide 98.0+%, TCI America™

Catalog No. D39765G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D39765G Supplier TCI America Supplier No. D39765G
Only null left

Chemical Identifiers

CAS 138666-59-8
Molecular Formula C9H16Br3N2-
Molecular Weight (g/mol) 391.953
MDL Number MFCD00191982
InChI Key DPWBUJYBOOWMLB-UHFFFAOYSA-N
Synonym 1,8-diazabicyclo 5.4.0 undec-7-ene hydrotribromide
PubChem CID 131864268
SMILES C1CCC2=NCCCN2CC1.Br[Br-]Br

Specifications

Melting Point 123°C
Quantity 5 g
UN Number 3261
Formula Weight 392.96
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Diazabicyclo[5.4.0]-7-undecene Hydrogen Tribromide