CAS RN 138666-59-8
CAS RN 138666-59-8
IUPAC Name:
2H,3H,4H,6H,7H,8H,9H,10H-pyrimido[1,2-a]azepine; tribroman-2-uide
Synonyms:
1,8-diazabicycloundec-7-ene; tribroman-2-uide
Molecular Weight (g/mol):
391.95
Molecular Formula:
C9H16Br3N2
InChi Key:
DPWBUJYBOOWMLB-UHFFFAOYSA-N
SMILES:
Br[Br-]Br.C1CN=C2CCCCCN2C1
1
–
1
of
1
results
1
1,8-Diazabicyclo[5.4.0]-7-undecene Hydrogen Tribromide 98.0+%, TCI America™
CAS: 138666-59-8 Molecular Formula: C9H16Br3N2- Molecular Weight (g/mol): 391.953 MDL Number: MFCD00191982 InChI Key: DPWBUJYBOOWMLB-UHFFFAOYSA-N Synonym: 1,8-diazabicyclo 5.4.0 undec-7-ene hydrotribromide PubChem CID: 131864268 SMILES: C1CCC2=NCCCN2CC1.Br[Br-]Br