missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diaminooctane 98.0+%, TCI America™

Catalog No. p-7126036
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D010725G 25 g
D0107100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D010725G Supplier TCI America Supplier No. D010725G
Only null left

Chemical Identifiers

CAS 373-44-4
Molecular Formula C8H22Cl2N2
Molecular Weight (g/mol) 217.18
MDL Number MFCD00008248
InChI Key ZFLWZOGXFQNIMT-UHFFFAOYSA-N
Synonym 1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392
PubChem CID 24250
ChEBI CHEBI:73112
IUPAC Name octane-1,8-bis(aminium) dichloride
SMILES [Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+]

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 240°C
Quantity 25 g
UN Number 3259
Formula Weight 144.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Diaminooctane
EINECSNumber (2)-3108
RTECSNumber RG8841500
TSCA No