accessibility menu, dialog, popup

UserName

H2NNH2

CAS RN 373-44-4

IUPAC Name: octane-1,8-bis(aminium) dichloride
Synonyms: octane-1,8-bis(aminium) dichloride
Molecular Weight (g/mol): 217.18
Molecular Formula: C8H22Cl2N2
InChi Key: ZFLWZOGXFQNIMT-UHFFFAOYSA-N
SMILES: [Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+]
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (1)
Quantity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)