Learn More
1,8-Diaminonaphthalene, 97%
CAS: 479-27-6 | C10H10N2 | 158.204 g/mol
Supplier: Thermo Scientific Chemicals A1596630
Description
1,8-Diaminonaphthalene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,8-Diaminonaphthalene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and strong oxidizing agents.
Specifications
| 1,8-Diaminonaphthalene | |
| 61°C to 65°C | |
| 195°C (383°F) | |
| MFCD00004033 | |
| 742841 | |
| 1,8-diaminonaphthalene, 1,8-naphthalenediamine, 1,8-naphthylenediamine, unii-ika7029yh9, ccris 8398, 1,8-diaminonapthalene, 1,8-diamino naphthalene, 1,8-naphthalene diamine, acmc-209kb1, ksc236e9t | |
| YFOOEYJGMMJJLS-UHFFFAOYSA-N | |
| naphthalene-1,8-diamine | |
| 68067 | |
| 97% |
| 479-27-6 | |
| 205°C (12 mmHg) | |
| C10H10N2 | |
| 250 g | |
| 14,6372 | |
| slightly soluble in water. | |
| C1=CC2=C(C(=C1)N)C(=CC=C2)N | |
| 158.204 | |
| 158.2 |
Chemical Identifiers
| 479-27-6 | |
| 158.204 | |
| YFOOEYJGMMJJLS-UHFFFAOYSA-N | |
| 68067 | |
| C1=CC2=C(C(=C1)N)C(=CC=C2)N |
| C10H10N2 | |
| MFCD00004033 | |
| 1,8-diaminonaphthalene, 1,8-naphthalenediamine, 1,8-naphthylenediamine, unii-ika7029yh9, ccris 8398, 1,8-diaminonapthalene, 1,8-diamino naphthalene, 1,8-naphthalene diamine, acmc-209kb1, ksc236e9t | |
| naphthalene-1,8-diamine |
Safety and Handling
GHS H Statement
H301-H351-H317
Toxic if swallowed.
Suspected of causing cancer.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
H302-H317
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 207-529-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only