missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diaminonaphthalene, 97%

CAS: 479-27-6 | C10H10N2 | 158.204 g/mol

Supplier:  Thermo Scientific Chemicals A159660B

 View more versions of this product

Catalog No. AAA159660B


Only null left
Explore available promotions

Description

Description

1,8-Diaminonaphthalene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diaminonaphthalene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and strong oxidizing agents.
Specifications

Specifications

1,8-Diaminonaphthalene
61°C to 65°C
195°C (383°F)
MFCD00004033
742841
1,8-diaminonaphthalene, 1,8-naphthalenediamine, 1,8-naphthylenediamine, unii-ika7029yh9, ccris 8398, 1,8-diaminonapthalene, 1,8-diamino naphthalene, 1,8-naphthalene diamine, acmc-209kb1, ksc236e9t
YFOOEYJGMMJJLS-UHFFFAOYSA-N
naphthalene-1,8-diamine
68067
97%
479-27-6
205°C (12 mmHg)
C10H10N2
1000 g
14,6372
slightly soluble in water.
C1=CC2=C(C(=C1)N)C(=CC=C2)N
158.204
158.2

Chemical Identifiers

479-27-6
158.204
YFOOEYJGMMJJLS-UHFFFAOYSA-N
68067
C1=CC2=C(C(=C1)N)C(=CC=C2)N
C10H10N2
MFCD00004033
1,8-diaminonaphthalene, 1,8-naphthalenediamine, 1,8-naphthylenediamine, unii-ika7029yh9, ccris 8398, 1,8-diaminonapthalene, 1,8-diamino naphthalene, 1,8-naphthalene diamine, acmc-209kb1, ksc236e9t
naphthalene-1,8-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H351-H317
Toxic if swallowed.
Suspected of causing cancer.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 207-529-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only