missing translation for 'onlineSavingsMsg'
Learn More

1,8-Cineole 98.0+%, TCI America™

Catalog No. C093425ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C093425ML 25 mL
C0934100ML 100 mL
C0934500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C093425ML Supplier TCI America Supplier No. C093425ML

Chemical Identifiers

CAS 470-82-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00167977
InChI Key WEEGYLXZBRQIMU-UHFFFAOYSA-N
Synonym eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan
PubChem CID 2758
IUPAC Name 1,3,3-trimethyl-2-oxabicyclo[2.2.2]octane
SMILES CC12CCC(CC1)C(C)(C)O2

Specifications

Melting Point 2°C
Color Colorless
Boiling Point 176°C
Quantity 25 mL
UN Number 1993
Formula Weight 154.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,8-Cineole
EINECSNumber (5)-0684
RTECSNumber OS9275000
TSCA Yes