Learn More
1,8-Cineole, 99%
CAS: 470-82-6 | C10H18O | 154.25 g/mol
Supplier: Thermo Scientific Chemicals 110341000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1,8-Cineole | |
| 1.5°C | |
| 0.9225g/mL | |
| 49°C | |
| 98.5% min. (GC) | |
| C10H18O | |
| MFCD00167977 | |
| 17,II,32 | |
| 15, 3938 | |
| eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan | |
| WEEGYLXZBRQIMU-UHFFFAOYSA-N | |
| 2,2,4-trimethyl-3-oxabicyclo[2.2.2]octane | |
| 2758 | |
| 99% |
| 470-82-6 | |
| Colorless to Yellow | |
| 176°C to 177°C | |
| Authentic | |
| Glass bottle | |
| 1.4550 to 1.46 | |
| 100 g | |
| 0.9225 | |
| -0.3° to +0.3° (20°C, 589nm) (neat) | |
| (in ethanol (70% v/v)) 1:2 | |
| CC12CCC(CC1)C(C)(C)O2 | |
| 154.25 | |
| 154.25 | |
| Liquid |
Chemical Identifiers
| 470-82-6 | |
| 154.25 | |
| WEEGYLXZBRQIMU-UHFFFAOYSA-N | |
| 2758 | |
| CC12CCC(CC1)C(C)(C)O2 |
| C10H18O | |
| MFCD00167977 | |
| eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan | |
| 2,2,4-trimethyl-3-oxabicyclo[2.2.2]octane |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May cause an allergic skin reaction.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Warning
EINECSNumber : 207-431-5
RUO – Research Use Only