missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Bis(dimethylamino)naphthalene 98.0+%, TCI America™
$55.92 - $305.70
Chemical Identifiers
| CAS | 20734-58-1 |
|---|---|
| Molecular Formula | C14H18N2 |
| Molecular Weight (g/mol) | 214.312 |
| MDL Number | MFCD00003920 |
| InChI Key | GJFNRSDCSTVPCJ-UHFFFAOYSA-N |
| Synonym | 1,8-bis dimethylamino naphthalene, n1,n1,n8,n8-tetramethylnaphthalene-1,8-diamine, proton sponge, 1,8-bis dimethylamino naphtalene, n,n,n',n'-tetramethyl-1,8-naphthalenediamine, proton-sponge, 1,8-naphthalenediamine, n,n,n',n'-tetramethyl, unii-6s79d2p9h8, 1,8-bis-dimethylamino naphthalene, n,n,n',n'-tetramethylnaphthalene-1,8-diamine |
| PubChem CID | 88675 |
| IUPAC Name | 1-N,1-N,8-N,8-N-tetramethylnaphthalene-1,8-diamine |
| SMILES | CN(C)C1=CC=CC2=C1C(=CC=C2)N(C)C |
Chemical Identifiers
| 20734-58-1 | |
| 214.312 | |
| GJFNRSDCSTVPCJ-UHFFFAOYSA-N | |
| 88675 | |
| CN(C)C1=CC=CC2=C1C(=CC=C2)N(C)C |
| C14H18N2 | |
| MFCD00003920 | |
| 1,8-bis dimethylamino naphthalene, n1,n1,n8,n8-tetramethylnaphthalene-1,8-diamine, proton sponge, 1,8-bis dimethylamino naphtalene, n,n,n',n'-tetramethyl-1,8-naphthalenediamine, proton-sponge, 1,8-naphthalenediamine, n,n,n',n'-tetramethyl, unii-6s79d2p9h8, 1,8-bis-dimethylamino naphthalene, n,n,n',n'-tetramethylnaphthalene-1,8-diamine | |
| 1-N,1-N,8-N,8-N-tetramethylnaphthalene-1,8-diamine |
Specifications
| 20734-58-1 | |
| 145°C | |
| MFCD00003920 | |
| 1,8-bis dimethylamino naphthalene, n1,n1,n8,n8-tetramethylnaphthalene-1,8-diamine, proton sponge, 1,8-bis dimethylamino naphtalene, n,n,n',n'-tetramethyl-1,8-naphthalenediamine, proton-sponge, 1,8-naphthalenediamine, n,n,n',n'-tetramethyl, unii-6s79d2p9h8, 1,8-bis-dimethylamino naphthalene, n,n,n',n'-tetramethylnaphthalene-1,8-diamine | |
| CN(C)C1=CC=CC2=C1C(=CC=C2)N(C)C | |
| 214.312 | |
| 214.31 | |
| Crystalline Powder |
| 48°C | |
| C14H18N2 | |
| 1 g | |
| GJFNRSDCSTVPCJ-UHFFFAOYSA-N | |
| 1-N,1-N,8-N,8-N-tetramethylnaphthalene-1,8-diamine | |
| 88675 | |
| ≥98.0% (GC,T) | |
| 1,8-Bis(dimethylamino)naphthalene [for Dehydrohalogenation] |
Safety and Handling
TSCA : No