missing translation for 'onlineSavingsMsg'
Learn More

1,8-Bis(dimethylamino)naphthalene 98.0+%, TCI America™

Catalog No. B10181G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10181G 1 g
B10185G 5 g
B101825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B10181G Supplier TCI America Supplier No. B10181G

Chemical Identifiers

CAS 20734-58-1
Molecular Formula C14H18N2
Molecular Weight (g/mol) 214.312
MDL Number MFCD00003920
InChI Key GJFNRSDCSTVPCJ-UHFFFAOYSA-N
Synonym 1,8-bis dimethylamino naphthalene, n1,n1,n8,n8-tetramethylnaphthalene-1,8-diamine, proton sponge, 1,8-bis dimethylamino naphtalene, n,n,n',n'-tetramethyl-1,8-naphthalenediamine, proton-sponge, 1,8-naphthalenediamine, n,n,n',n'-tetramethyl, unii-6s79d2p9h8, 1,8-bis-dimethylamino naphthalene, n,n,n',n'-tetramethylnaphthalene-1,8-diamine
PubChem CID 88675
IUPAC Name 1-N,1-N,8-N,8-N-tetramethylnaphthalene-1,8-diamine
SMILES CN(C)C1=CC=CC2=C1C(=CC=C2)N(C)C

Specifications

Melting Point 48°C
Boiling Point 145°C
Quantity 1 g
Formula Weight 214.31
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Bis(dimethylamino)naphthalene [for Dehydrohalogenation]