Learn More
1,8-Bis(dimethylamino)naphthalene, 98+%
CAS: 20734-58-1 | C14H18N2 | 214.312 g/mol
$78.73 - $285.99
Chemical Identifiers
| CAS | 20734-58-1 |
|---|---|
| Molecular Formula | C14H18N2 |
| Molecular Weight (g/mol) | 214.312 |
| MDL Number | MFCD00003920 |
| InChI Key | GJFNRSDCSTVPCJ-UHFFFAOYSA-N |
| Synonym | 1,8-bis dimethylamino naphthalene, n1,n1,n8,n8-tetramethylnaphthalene-1,8-diamine, proton sponge, 1,8-bis dimethylamino naphtalene, n,n,n',n'-tetramethyl-1,8-naphthalenediamine, proton-sponge, 1,8-naphthalenediamine, n,n,n',n'-tetramethyl, unii-6s79d2p9h8, 1,8-bis-dimethylamino naphthalene, n,n,n',n'-tetramethylnaphthalene-1,8-diamine |
| PubChem CID | 88675 |
| IUPAC Name | 1-N,1-N,8-N,8-N-tetramethylnaphthalene-1,8-diamine |
| SMILES | CN(C)C1=CC=CC2=C1C(=CC=C2)N(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0031309
|
Thermo Scientific Chemicals
L0031309 |
10 g |
Each for $78.73
|
|
|||||
|
AAL0031318
|
Thermo Scientific Chemicals
L0031318 |
50 g |
Each for $285.99
|
|
|||||
Description
1,8-Bis(dimethylamino)naphthalene is used in the synthesis of C-substituted t-BuNH-8,9-R,R?-nido-7,8,9-C3B8H9 (R,R? = H,H; MeH; Me,Me; Ph,H and Ph,Ph) tricarbollide compounds. It was also used in the preparation of saturated fluoroalkyl(hydrido) complexes of Iridium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,8-Bis(dimethylamino)naphthalene is used in the synthesis of C-substituted t-BuNH-8,9-R,R′-nido-7,8,9-C3B8H9 (R,R′ = H,H; MeH; Me,Me; Ph,H and Ph,Ph) tricarbollide compounds. It was also used in the preparation of saturated fluoroalkyl(hydrido) complexes of Iridium.
Solubility
Soluble in organic solvents.
Notes
Air & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 20734-58-1 | |
| 214.312 | |
| GJFNRSDCSTVPCJ-UHFFFAOYSA-N | |
| 88675 | |
| CN(C)C1=CC=CC2=C1C(=CC=C2)N(C)C |
| C14H18N2 | |
| MFCD00003920 | |
| 1,8-bis dimethylamino naphthalene, n1,n1,n8,n8-tetramethylnaphthalene-1,8-diamine, proton sponge, 1,8-bis dimethylamino naphtalene, n,n,n',n'-tetramethyl-1,8-naphthalenediamine, proton-sponge, 1,8-naphthalenediamine, n,n,n',n'-tetramethyl, unii-6s79d2p9h8, 1,8-bis-dimethylamino naphthalene, n,n,n',n'-tetramethylnaphthalene-1,8-diamine | |
| 1-N,1-N,8-N,8-N-tetramethylnaphthalene-1,8-diamine |
Specifications
| 20734-58-1 | |
| 144°C to 145°C (7 mmHg) | |
| C14H18N2 | |
| 10 g | |
| 396782 | |
| 1,8-bis dimethylamino naphthalene, n1,n1,n8,n8-tetramethylnaphthalene-1,8-diamine, proton sponge, 1,8-bis dimethylamino naphtalene, n,n,n',n'-tetramethyl-1,8-naphthalenediamine, proton-sponge, 1,8-naphthalenediamine, n,n,n',n'-tetramethyl, unii-6s79d2p9h8, 1,8-bis-dimethylamino naphthalene, n,n,n',n'-tetramethylnaphthalene-1,8-diamine | |
| GJFNRSDCSTVPCJ-UHFFFAOYSA-N | |
| 1-N,1-N,8-N,8-N-tetramethylnaphthalene-1,8-diamine | |
| 88675 | |
| ≥98% |
| 47°C to 51°C | |
| >110°C (230°F) | |
| MFCD00003920 | |
| UN3263 | |
| Air and light sensitive | |
| Soluble in organic solvents. | |
| CN(C)C1=CC=CC2=C1C(=CC=C2)N(C)C | |
| 214.312 | |
| 214.31 | |
| 1,8-Bis(dimethylamino)naphthalene |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
EINECSNumber : 244-001-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only