Learn More
1,8,9-Trihydroxyanthracene, 97%
CAS: 1143-38-0 | C14H10O3 | 226.23 g/mol
Supplier: Thermo Scientific Chemicals 457770050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1,8,9-Trihydroxyanthracene | |
| 178.0°C to 181.0°C | |
| 96% min. (HPLC) | |
| MFCD00053409 | |
| anthralin, dithranol, 1,8-dihydroxyanthrone, 1,8-dihydroxy-9-anthrone, chrysodermol, cigthranol, batridol, cignolin, psoriacid-stift, 9 10h-anthracenone, 1,8-dihydroxy | |
| NUZWLKWWNNJHPT-UHFFFAOYSA-N | |
| 1,8-dihydroxy-10H-anthracen-9-one | |
| 2202 | |
| 226.23 |
| 1143-38-0 | |
| Authentic | |
| C14H10O3 | |
| 5 g | |
| Solubility in water: insoluble. Other solubilities: soluble in chloroform, acetone, benzene, slightly soluble in alcohol, ether, glacial acetic | |
| OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2 | |
| 226.23 | |
| CHEBI:37510 | |
| 97% |
Chemical Identifiers
| 1143-38-0 | |
| 226.23 | |
| NUZWLKWWNNJHPT-UHFFFAOYSA-N | |
| 2202 | |
| 1,8-dihydroxy-10H-anthracen-9-one |
| C14H10O3 | |
| MFCD00053409 | |
| anthralin, dithranol, 1,8-dihydroxyanthrone, 1,8-dihydroxy-9-anthrone, chrysodermol, cigthranol, batridol, cignolin, psoriacid-stift, 9 10h-anthracenone, 1,8-dihydroxy | |
| CHEBI:37510 | |
| OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 214-538-
RUO – Research Use Only