missing translation for 'onlineSavingsMsg'
Learn More

1,8,9-Trihydroxyanthracene, 97%

Catalog No. AC457770010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457770010 1 g
AC457770050 5 g
AC457770250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC457770010 Supplier Thermo Scientific Chemicals Supplier No. 457770010

CAS: 1143-38-0 | C14H10O3 | 226.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1143-38-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00053409
InChI Key NUZWLKWWNNJHPT-UHFFFAOYSA-N
Synonym anthralin, dithranol, 1,8-dihydroxyanthrone, 1,8-dihydroxy-9-anthrone, chrysodermol, cigthranol, batridol, cignolin, psoriacid-stift, 9 10h-anthracenone, 1,8-dihydroxy
PubChem CID 2202
ChEBI CHEBI:37510
IUPAC Name 1,8-dihydroxy-10H-anthracen-9-one
SMILES OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2

Specifications

Melting Point 178.0°C to 181.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, acetone, benzene, slightly soluble in alcohol, ether, glacial acetic
Formula Weight 226.23
Percent Purity 97%
Chemical Name or Material 1,8,9-Trihydroxyanthracene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 214-538-

RUO – Research Use Only