missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,7-Octadiyne 98.0+%, TCI America™
$106.25 - $303.84
Chemical Identifiers
| CAS | 871-84-1 |
|---|---|
| Molecular Formula | C8H10 |
| Molecular Weight (g/mol) | 106.17 |
| MDL Number | MFCD00008580 |
| InChI Key | DSOJWVLXZNRKCS-UHFFFAOYSA-N |
| Synonym | 1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ci |
| PubChem CID | 70099 |
| IUPAC Name | octa-1,7-diyne |
| SMILES | C#CCCCCC#C |
Chemical Identifiers
| 871-84-1 | |
| 106.17 | |
| DSOJWVLXZNRKCS-UHFFFAOYSA-N | |
| 70099 | |
| C#CCCCCC#C |
| C8H10 | |
| MFCD00008580 | |
| 1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ci | |
| octa-1,7-diyne |
Specifications
| 871-84-1 | |
| 135°C | |
| MFCD00008580 | |
| 3295 | |
| DSOJWVLXZNRKCS-UHFFFAOYSA-N | |
| octa-1,7-diyne | |
| 70099 | |
| ≥98.0% (GC) | |
| 1,7-Octadiyne |
| Yellow | |
| C8H10 | |
| 5 mL | |
| 1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ci | |
| C#CCCCCC#C | |
| 106.17 | |
| 106.17 | |
| Liquid |
Safety and Handling
TSCA : Yes