Learn More
1,7-Octadiyne, 98%
CAS: 871-84-1 | C8H10 | 106.17 g/mol
$171.37 - $551.63
Chemical Identifiers
| CAS | 871-84-1 |
|---|---|
| Molecular Formula | C8H10 |
| Molecular Weight (g/mol) | 106.17 |
| MDL Number | MFCD00008580 |
| InChI Key | DSOJWVLXZNRKCS-UHFFFAOYSA-N |
| Synonym | 1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ci |
| PubChem CID | 70099 |
| IUPAC Name | octa-1,7-diyne |
| SMILES | C#CCCCCC#C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC171340100
|
Thermo Scientific Chemicals
171340100 |
10 g | Glass bottle |
Each for $171.37
|
|
||||
|
AC171340500
|
Thermo Scientific Chemicals
171340500 |
50 g | Glass bottle |
Each for $551.63
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 871-84-1 | |
| 106.17 | |
| DSOJWVLXZNRKCS-UHFFFAOYSA-N | |
| 70099 | |
| C#CCCCCC#C |
| C8H10 | |
| MFCD00008580 | |
| 1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ci | |
| octa-1,7-diyne |
Specifications
| 871-84-1 | |
| 100.0 | |
| 0.8000g/mL | |
| 23°C | |
| 97.5% min. (GC) | |
| C8H10 | |
| HC≡C(CH2)4C≡CH | |
| 10 g | |
| 0.8 | |
| DSOJWVLXZNRKCS-UHFFFAOYSA-N | |
| octa-1,7-diyne | |
| 70099 | |
| 98% | |
| 1, 7-Octadiyne, 98% |
| 98.5 | |
| Colorless to Yellow | |
| 135.0°C to 136.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4454 to 1.4474 | |
| MFCD00008580 | |
| 01, II, 248 | |
| 1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ci | |
| C#CCCCCC#C | |
| 106.17 | |
| 106.16 | |
| Liquid |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 212-815-0
TSCA : TSCA
RUO – Research Use Only