Learn More
1,7-Octadiyne, 98%
CAS: 871-84-1 | C8H10 | 106.17 g/mol
$175.48 - $612.56
Chemical Identifiers
| CAS | 871-84-1 |
|---|---|
| Molecular Formula | C8H10 |
| Molecular Weight (g/mol) | 106.17 |
| MDL Number | MFCD00008580 |
| InChI Key | DSOJWVLXZNRKCS-UHFFFAOYSA-N |
| Synonym | 1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ci |
| PubChem CID | 70099 |
| IUPAC Name | octa-1,7-diyne |
| SMILES | C#CCCCCC#C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1582909
|
Thermo Scientific Chemicals
A1582909 |
10 g |
Each for $175.48
|
|
|||||
|
AAA1582918
|
Thermo Scientific Chemicals
A1582918 |
50 g |
Each for $612.56
|
|
|||||
Description
1,7-Octadiyne is an useful bifunctional chain extension unit. It participates in the formation of uranium(IV) vinyl complexes. It reacts with frustrated Lewis pair P(o-tolyl)3/B(C6F5)3 by acetylene C-C coupling to yield the zwitterionic product. It undergoes the reductive cyclization during reaction with Re2Cl4(?-dppm)2 (dppm = Ph2PCH2PPh2) and affords quadruply bonded dirhenium(III) complex Re2Cl3(?,η2-C8H7)(?-dppm)2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,7-Octadiyne is an useful bifunctional chain extension unit. It participates in the formation of uranium(IV) vinyl complexes. It reacts with frustrated Lewis pair P(o-tolyl)3/B(C6F5)3 by acetylene C-C coupling to yield the zwitterionic product. It undergoes the reductive cyclization during reaction with Re2Cl4(μ-dppm)2 (dppm = Ph2PCH2PPh2) and affords quadruply bonded dirhenium(III) complex Re2Cl3(μ,η2-C8H7)(μ-dppm)2.
Solubility
Not miscible or difficult to mix.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 871-84-1 | |
| 106.17 | |
| DSOJWVLXZNRKCS-UHFFFAOYSA-N | |
| 70099 | |
| C#CCCCCC#C |
| C8H10 | |
| MFCD00008580 | |
| 1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ci | |
| octa-1,7-diyne |
Specifications
| 871-84-1 | |
| 135°C to 136°C | |
| Recognizable | |
| 1.446 | |
| 10 g | |
| 1697275 | |
| Not miscible or difficult to mix. | |
| C#CCCCCC#C | |
| 106.17 | |
| 106.17 | |
| 1,7-Octadiyne |
| 0.81 | |
| 23°C (73°F) | |
| C8H10 | |
| MFCD00008580 | |
| UN3295 | |
| 1,7-octadiyne, 1,7 octadiyne, oct-1,7-diyne, acmc-209qii, 3-thia-1,5-pentanediamine, 1,7-octadiyne 10g, 1,7-octadiyne 6ci, 7ci, 8ci, 9ci | |
| DSOJWVLXZNRKCS-UHFFFAOYSA-N | |
| octa-1,7-diyne | |
| 70099 | |
| 98% |
Safety and Handling
GHS H Statement
H304-H226-H315-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 212-815-0
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only