missing translation for 'onlineSavingsMsg'
Learn More

1,7-Dichloro-4-methoxyisoquinoline 98.0+%, TCI America™

Catalog No. D51821G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D51821G Supplier TCI America Supplier No. D51821G
Only null left

Chemical Identifiers

CAS 630423-36-8
Molecular Formula C10H7Cl2NO
Molecular Weight (g/mol) 228.072
MDL Number MFCD17012717
InChI Key HOBCGPSQZNVUSJ-UHFFFAOYSA-N
PubChem CID 22250249
IUPAC Name 1,7-dichloro-4-methoxyisoquinoline
SMILES COC1=CN=C(C2=C1C=CC(=C2)Cl)Cl

Specifications

Melting Point 139°C
Quantity 1 g
Formula Weight 228.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,7-Dichloro-4-methoxyisoquinoline