accessibility menu, dialog, popup

UserName

H3CONClCl

CAS RN 630423-36-8

IUPAC Name: 1,7-dichloro-4-methoxyisoquinoline
Synonyms: 1,7-dichloro-4-methoxyisoquinoline
Molecular Weight (g/mol): 228.07
Molecular Formula: C10H7Cl2NO
InChi Key: HOBCGPSQZNVUSJ-UHFFFAOYSA-N
SMILES: COC1=C2C=CC(Cl)=CC2=C(Cl)N=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results