missing translation for 'onlineSavingsMsg'
Learn More

1,7-Dibromo-3,4,9,10-perylenetetracarboxylic Dianhydride 98.0+%, TCI America™

Catalog No. p-7127966
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38711G 1 g
D38715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38711G Supplier TCI America Supplier No. D38711G
Only null left

Chemical Identifiers

CAS 118129-60-5
Molecular Formula C24H6Br2O6
Molecular Weight (g/mol) 550.114
MDL Number MFCD11870875
InChI Key WPBVUAVIGWNDGT-UHFFFAOYSA-N
PubChem CID 11103604
SMILES C1=CC2=C3C(=CC(=C4C3=C1C5=C(C=C6C7=C(C=CC4=C57)C(=O)OC6=O)Br)Br)C(=O)OC2=O

Specifications

Melting Point 360°C
Quantity 1 g
Formula Weight 550.11
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,7-Dibromo-3,4,9,10-perylenetetracarboxylic Dianhydride