accessibility menu, dialog, popup

UserName

CAS RN 118129-60-5

IUPAC Name: 11,22-dibromo-7,18-dioxaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Synonyms: 11,22-dibromo-7,18-dioxaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Molecular Weight (g/mol): 550.11
Molecular Formula: C24H6Br2O6
InChi Key: WPBVUAVIGWNDGT-UHFFFAOYSA-N
SMILES: BrC1=C2C3=CC=C4C(=O)OC(=O)C5=CC(Br)=C(C6=CC=C7C(=O)OC(=O)C(=C1)C7=C26)C3=C45
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results