missing translation for 'onlineSavingsMsg'
Learn More

1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol 98.0+%, TCI America™

Catalog No. p-7134107
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10691G 1 g
N10695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N10691G Supplier TCI America Supplier No. N10691G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 159873-64-0
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.173
MDL Number MFCD08274663
InChI Key UHJMDARCOIYCHL-UHFFFAOYSA-N
Synonym alpha-Methyl-6-nitro-1,3-benzodioxole-5-methanol, alpha-Methyl-6-nitropiperonyl Alcohol
PubChem CID 10932738
IUPAC Name 1-(6-nitro-1,3-benzodioxol-5-yl)ethanol
SMILES CC(C1=CC2=C(C=C1[N+](=O)[O-])OCO2)O

Specifications

Melting Point 88°C
Quantity 1 g
Formula Weight 211.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol