accessibility menu, dialog, popup

UserName

CH3NOOOOOH

CAS RN 159873-64-0

IUPAC Name: 1-(6-nitro-2H-1,3-benzodioxol-5-yl)ethan-1-ol
Synonyms: 1-(6-nitro-2H-1,3-benzodioxol-5-yl)ethanol
Molecular Weight (g/mol): 211.17
Molecular Formula: C9H9NO5
InChi Key: UHJMDARCOIYCHL-UHFFFAOYNA-N
SMILES: CC(O)C1=CC2=C(OCO2)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results