missing translation for 'onlineSavingsMsg'
Learn More

1,6-Naphthyridine 97.0+%, TCI America™

Catalog No. N0462100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0462100MG 100 mg
N04621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0462100MG Supplier TCI America Supplier No. N0462100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 253-72-5
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00059750
InChI Key VSOSXKMEQPYESP-UHFFFAOYSA-N
Synonym 1,6-diazanaphthalene, 1,6 naphthyridine, 1,6-pyridopyridine, unii-s1t14p7ow7, pyridino 4,3-b pyridine, 1,6naphthyridine, 1,6-naphthyridine, pubchem18071, acmc-1chvm, pyrido 4,3-b pyridine
PubChem CID 67488
ChEBI CHEBI:36627
IUPAC Name 1,6-naphthyridine
SMILES C1=CN=C2C=CN=CC2=C1

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 130.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Naphthyridine