accessibility menu, dialog, popup

UserName

CAS RN 253-72-5

IUPAC Name: 1,6-naphthyridine
Synonyms: 1,6-naphthyridine 1,6-pyridopyridine
Molecular Weight (g/mol): 130.15
Molecular Formula: C8H6N2
InChi Key: VSOSXKMEQPYESP-UHFFFAOYSA-N
SMILES: C1=CN=C2C=CN=CC2=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)