missing translation for 'onlineSavingsMsg'
Learn More

1,6-Naphthyridine, 96%, Thermo Scientific™

Catalog No. AC457670010
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457670010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC457670010 Supplier Thermo Scientific Chemicals Supplier No. 457670010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 253-72-5
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00059750
InChI Key VSOSXKMEQPYESP-UHFFFAOYSA-N
Synonym 1,6-diazanaphthalene, 1,6 naphthyridine, 1,6-pyridopyridine, unii-s1t14p7ow7, pyridino 4,3-b pyridine, 1,6naphthyridine, 1,6-naphthyridine, pubchem18071, acmc-1chvm, pyrido 4,3-b pyridine
PubChem CID 67488
ChEBI CHEBI:36627
IUPAC Name 1,6-naphthyridine
SMILES C1=CN=C2C=CN=CC2=C1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Quantity 1g
Formula Weight 130.15
Percent Purity 96%
Chemical Name or Material 1,6-Naphthyridine

RUO – Research Use Only