missing translation for 'onlineSavingsMsg'
Learn More

1,6-Naphthyridin-2(1H)-one, 97%, Thermo Scientific Chemicals

$457.22 - $457.22

Chemical Identifiers

CAS 23616-29-7
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.15
MDL Number MFCD07781179
InChI Key ZQKMVHXJWJNEQG-UHFFFAOYSA-N
Synonym 1,6-naphthyridin-2 1h-one, acmc-1cgmu, pyrido 3,2-c pyridone, hydroxy-1,6-naphthyridine, 1,6-naphthyridine-2-one, 2-hydroxy-1,6-naphthyridine, 1,6 naphthyridin-2-ol #, 1h-1,6 naphthyridin-2-one
PubChem CID 589674
IUPAC Name 1H-1,6-naphthyridin-2-one
SMILES O=C1NC2=CC=NC=C2C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5012103
View Documents
Thermo Scientific Chemicals
H5012103
1 g
Each for $457.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

23616-29-7
146.15
ZQKMVHXJWJNEQG-UHFFFAOYSA-N
589674
O=C1NC2=CC=NC=C2C=C1
C8H6N2O
MFCD07781179
1,6-naphthyridin-2 1h-one, acmc-1cgmu, pyrido 3,2-c pyridone, hydroxy-1,6-naphthyridine, 1,6-naphthyridine-2-one, 2-hydroxy-1,6-naphthyridine, 1,6 naphthyridin-2-ol #, 1h-1,6 naphthyridin-2-one
1H-1,6-naphthyridin-2-one

Specifications

23616-29-7
MFCD07781179
Air Sensitive
ZQKMVHXJWJNEQG-UHFFFAOYSA-N
1H-1,6-naphthyridin-2-one
589674
97%
C8H6N2O
1 g
1,6-naphthyridin-2 1h-one, acmc-1cgmu, pyrido 3,2-c pyridone, hydroxy-1,6-naphthyridine, 1,6-naphthyridine-2-one, 2-hydroxy-1,6-naphthyridine, 1,6 naphthyridin-2-ol #, 1h-1,6 naphthyridin-2-one
O=C1NC2=CC=NC=C2C=C1
146.15
146.15
1,6-Naphthyridin-2(1H)-one
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only