accessibility menu, dialog, popup

UserName

CAS RN 23616-29-7

ONHN

CAS RN 23616-29-7

IUPAC Name: 1,2-dihydro-1,6-naphthyridin-2-one
Synonyms: 1H-1,6-naphthyridin-2-one
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: ZQKMVHXJWJNEQG-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=NC=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results