missing translation for 'onlineSavingsMsg'
Learn More

[1-(6-Methylpyrazin-2-yl)piperid-4-yl]methylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 892502-20-4
Molecular Formula C11H18N4
Molecular Weight (g/mol) 206.293
MDL Number MFCD09025897
InChI Key XVEZCDMRMDBGQU-UHFFFAOYSA-N
Synonym 1-6-methylpyrazin-2-yl piperid-4-yl methylamine, 1-6-methylpyrazin-2-yl piperidin-4-yl methanamine, 1-1-6-methylpyrazin-2-yl piperidin-4-yl methanamine, 1-6-methylpyrazin-2-yl-4-piperidyl methylamine
PubChem CID 18525899
IUPAC Name [1-(6-methylpyrazin-2-yl)piperidin-4-yl]methanamine
SMILES CC1=CN=CC(=N1)N2CCC(CC2)CN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC58913CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC58913DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

892502-20-4
206.293
XVEZCDMRMDBGQU-UHFFFAOYSA-N
18525899
CC1=CN=CC(=N1)N2CCC(CC2)CN
C11H18N4
MFCD09025897
1-6-methylpyrazin-2-yl piperid-4-yl methylamine, 1-6-methylpyrazin-2-yl piperidin-4-yl methanamine, 1-1-6-methylpyrazin-2-yl piperidin-4-yl methanamine, 1-6-methylpyrazin-2-yl-4-piperidyl methylamine
[1-(6-methylpyrazin-2-yl)piperidin-4-yl]methanamine

Specifications

892502-20-4
100.0
C11H18N4
1-6-methylpyrazin-2-yl piperid-4-yl methylamine, 1-6-methylpyrazin-2-yl piperidin-4-yl methanamine, 1-1-6-methylpyrazin-2-yl piperidin-4-yl methanamine, 1-6-methylpyrazin-2-yl-4-piperidyl methylamine
CC1=CN=CC(=N1)N2CCC(CC2)CN
206.293
206.29
[1-(6-Methylpyrazin-2-yl)piperid-4-yl]methylamine
96.5
97%
MFCD09025897
XVEZCDMRMDBGQU-UHFFFAOYSA-N
[1-(6-methylpyrazin-2-yl)piperidin-4-yl]methanamine
18525899
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents