missing translation for 'onlineSavingsMsg'
Learn More

[1-(6-Methylpyrazin-2-yl)piperid-4-yl]methylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC58913DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC58913DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

892502-20-4
206.293
XVEZCDMRMDBGQU-UHFFFAOYSA-N
18525899
CC1=CN=CC(=N1)N2CCC(CC2)CN
C11H18N4
MFCD09025897
1-6-methylpyrazin-2-yl piperid-4-yl methylamine, 1-6-methylpyrazin-2-yl piperidin-4-yl methanamine, 1-1-6-methylpyrazin-2-yl piperidin-4-yl methanamine, 1-6-methylpyrazin-2-yl-4-piperidyl methylamine
[1-(6-methylpyrazin-2-yl)piperidin-4-yl]methanamine

Specifications

[1-(6-Methylpyrazin-2-yl)piperid-4-yl]methylamine
96.5
97%
C11H18N4
MFCD09025897
XVEZCDMRMDBGQU-UHFFFAOYSA-N
[1-(6-methylpyrazin-2-yl)piperidin-4-yl]methanamine
18525899
97%
892502-20-4
100.0
Amber Glass Bottle
1g
1-6-methylpyrazin-2-yl piperid-4-yl methylamine, 1-6-methylpyrazin-2-yl piperidin-4-yl methanamine, 1-1-6-methylpyrazin-2-yl piperidin-4-yl methanamine, 1-6-methylpyrazin-2-yl-4-piperidyl methylamine
CC1=CN=CC(=N1)N2CCC(CC2)CN
206.293
206.29
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents