missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanedithiol, 97%

CAS: 1191-43-1 | C6H14S2 | 150.30 g/mol

$53.84 - $121.45

Chemical Identifiers

CAS 1191-43-1
Molecular Formula C6H14S2
Molecular Weight (g/mol) 150.30
MDL Number MFCD00004910
InChI Key SRZXCOWFGPICGA-UHFFFAOYSA-N
Synonym 1,6-hexanedithiol, 1,6-dimercaptohexane, 1,6-hexanedimercaptan, usaf uctl-72, hexamethylene dimercaptan, 1,6-hexane dithiol, 1,6-hexamethylenedithiol, unii-rwn7rm884e, fema no. 3495, hexanedithiol-1,6
PubChem CID 14491
IUPAC Name hexane-1,6-dithiol
SMILES SCCCCCCS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0668606
View Documents
Thermo Scientific Chemicals
L0668606
5 g
Each for $53.84
Only null left
 
AAL0668614
View Documents
Thermo Scientific Chemicals
L0668614
25 g
Each for $121.45
Only null left
 
Description

Description

1,6-Hexanedithiol is used in self-assembled monolayers (SAMs). It is used in synthetic rubber and food with spices. It is also used in Food Flavor, Tobacco Flavor, Blending edible flavors: Beef, poultry soup .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Hexanedithiol is used in self-assembled monolayers (SAMs). It is used in synthetic rubber and food with spices. It is also used in Food Flavor, Tobacco Flavor, Blending edible flavors: Beef, poultry soup .

Solubility
Not miscible in water.

Notes
Incompatible materials are Bases, Oxidizing agents, Reducing agents, Alkali metals. Avoid Heat and direct sunlight.
Specifications

Chemical Identifiers

1191-43-1
150.30
SRZXCOWFGPICGA-UHFFFAOYSA-N
14491
SCCCCCCS
C6H14S2
MFCD00004910
1,6-hexanedithiol, 1,6-dimercaptohexane, 1,6-hexanedimercaptan, usaf uctl-72, hexamethylene dimercaptan, 1,6-hexane dithiol, 1,6-hexamethylenedithiol, unii-rwn7rm884e, fema no. 3495, hexanedithiol-1,6
hexane-1,6-dithiol

Specifications

1191-43-1
0.99
90°C (194°F)
1.513
5 g
1732507
Not miscible in water.
SCCCCCCS
150.30
150.3
1,6-Hexanedithiol
-21°C
242°C to 243°C
C6H14S2
MFCD00004910
UN2810
1,6-hexanedithiol, 1,6-dimercaptohexane, 1,6-hexanedimercaptan, usaf uctl-72, hexamethylene dimercaptan, 1,6-hexane dithiol, 1,6-hexamethylenedithiol, unii-rwn7rm884e, fema no. 3495, hexanedithiol-1,6
SRZXCOWFGPICGA-UHFFFAOYSA-N
hexane-1,6-dithiol
14491
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H227-H302-H312
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.

P210-P280-P370+P378q-P501c

H227

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 214-735-1

RTECSNumber : MO3500000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only