Learn More
1,6-Hexanedithiol, 97%
CAS: 1191-43-1 | C6H14S2 | 150.30 g/mol
Supplier: Thermo Scientific Chemicals L0668614
Description
1,6-Hexanedithiol is used in self-assembled monolayers (SAMs). It is used in synthetic rubber and food with spices. It is also used in Food Flavor, Tobacco Flavor, Blending edible flavors: Beef, poultry soup .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,6-Hexanedithiol is used in self-assembled monolayers (SAMs). It is used in synthetic rubber and food with spices. It is also used in Food Flavor, Tobacco Flavor, Blending edible flavors: Beef, poultry soup .
Solubility
Not miscible in water.
Notes
Incompatible materials are Bases, Oxidizing agents, Reducing agents, Alkali metals. Avoid Heat and direct sunlight.
Specifications
| 1,6-Hexanedithiol | |
| -21°C | |
| 242°C to 243°C | |
| C6H14S2 | |
| MFCD00004910 | |
| UN2810 | |
| 1,6-hexanedithiol, 1,6-dimercaptohexane, 1,6-hexanedimercaptan, usaf uctl-72, hexamethylene dimercaptan, 1,6-hexane dithiol, 1,6-hexamethylenedithiol, unii-rwn7rm884e, fema no. 3495, hexanedithiol-1,6 | |
| SRZXCOWFGPICGA-UHFFFAOYSA-N | |
| hexane-1,6-dithiol | |
| 14491 | |
| 97% |
| 1191-43-1 | |
| 0.99 | |
| 90°C (194°F) | |
| 1.513 | |
| 25 g | |
| 1732507 | |
| Not miscible in water. | |
| SCCCCCCS | |
| 150.30 | |
| 150.3 |
Chemical Identifiers
| 1191-43-1 | |
| 150.30 | |
| SRZXCOWFGPICGA-UHFFFAOYSA-N | |
| 14491 | |
| SCCCCCCS |
| C6H14S2 | |
| MFCD00004910 | |
| 1,6-hexanedithiol, 1,6-dimercaptohexane, 1,6-hexanedimercaptan, usaf uctl-72, hexamethylene dimercaptan, 1,6-hexane dithiol, 1,6-hexamethylenedithiol, unii-rwn7rm884e, fema no. 3495, hexanedithiol-1,6 | |
| hexane-1,6-dithiol |
Safety and Handling
GHS H Statement
H331-H227-H302-H312
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
P210-P280-P370+P378q-P501c
H227
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 214-735-1
RTECSNumber : MO3500000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only