missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediol Dimethacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. H136925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H136925G 25 g
H1369500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H136925G Supplier TCI America Supplier No. H136925G
Only null left

Chemical Identifiers

CAS 6606-59-3
Molecular Formula C14H22O4
Molecular Weight (g/mol) 254.326
MDL Number MFCD00015425
InChI Key SAPGBCWOQLHKKZ-UHFFFAOYSA-N
Synonym 1,6-hexanediol dimethacrylate, unii-rao49190jv, 1,6-hexanediyl bismethacrylate, ccris 7049, 1,6-hexamethylene dimethacrylate, 2-propenoic acid, 2-methyl-, 1,6-hexanediyl ester, hexane-1,6-diyl bis 2-methylacrylate, 2-propenoic acid, 2-methyl-, 1,1'-1,6-hexanediyl ester, 1,6-hexanediyl bis 2-methylacrylate, 6-2-methylprop-2-enoyl oxy hexyl 2-methylprop-2-enoate
PubChem CID 81080
IUPAC Name 6-(2-methylprop-2-enoyloxy)hexyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCCCCCOC(=O)C(=C)C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 254.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,6-Hexanediol Dimethacrylate (stabilized with MEHQ)
EINECSNumber (2)-0958&(2)-1059
TSCA Yes