missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediol diacrylate, 99% (reactive esters), stab. with 90ppm hydroquinone

Catalog No. AA4320330
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4320330 250 g
AA4320318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4320330 Supplier Thermo Scientific Chemicals Supplier No. 04320330

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13048-33-4 | C12H18O4 | 226.27 g/mol

1,6-Hexanediol diacrylate is used as a functional monomer for polymers. It acts as a cross linking agent between the molecular chains of polymers. Further, it is used in adhesives, sealants, alkyd coatings, elastomers, photopolymers, and inks for improved adhesion, hardness, abrasion and heat resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13048-33-4
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.27
MDL Number MFCD00008631
InChI Key FIHBHSQYSYVZQE-UHFFFAOYSA-N
Synonym 1,6-hexanediol diacrylate, hexamethylene diacrylate, kayarad hdda, hexane-1,6-diyl diacrylate, 1,6-hexamethylene diacrylate, hexamethylene glycol diacrylate, sartomer 238, hdoda, viscoat 230, hexamethylene acrylate
PubChem CID 25644
IUPAC Name 6-prop-2-enoyloxyhexyl prop-2-enoate
SMILES C=CC(=O)OCCCCCCOC(=O)C=C

Specifications

Density 1.010 g/mL
Flash Point >110°C (>230°F)
Quantity 250 g
Sensitivity Hygroscopic
Solubility Information Slightly miscible with water.
Formula Weight 226.28
Percent Purity 99%
Physical Form Liquid
Assay (reactive esters)
Chemical Name or Material 1,6-Hexanediol diacrylate, Stabilized with 90ppm hydroquinone
Hazard Category H315-H317-H319
Hazard Statement GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
EINECSNumber 235-921-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only